7007309 - Rev 1 - Horizontal Systems, Models B1A, B1, B2 and B3 - Operating and Maintenance Manual
Description:
Operating and maintenance manual
Environment:
Horizontal Minigel Systems - Models:
- B1A
- B1
- B2
Self Recirculation System - Model:
- B3
Specifications and recommended running conditions:
Model | B1A | B1 | B2 | B3 |
Gel Size (Wx L in cm.) | 7x8 | 9x11 | 12x14 | 12x14 |
Buffer Capacity | 400ml | 600ml | 800ml | 1000ml |
Voltage Requirements (V) | 20-150 | 20-150 | 20-150 | 20-150 |
Time Requirements (minutes) | 30-60 | 45-90 | 60-120 | 60-120 |
Attachment(s):
File | Last Modified |
---|---|
7007309 - Rev 1 - Horizontal Systems, Models B1A, B1, B2 and B3 - Operating and Maintenance Manual.pdf | December 15, 2021 |