7007320 - Rev 2 - Horizontal Electrophoresis System, Model D2 and D3-14 - Operating and Maintenance Manual
Description:
Operating and maintenance manual
Environment:
- Owl Model D3-14 Horizontal Agarose Gel Electrophoresis System
- Owl Model D2 Horizontal Agarose Gel Electrophoresis System
Recommended running conditions:
Model | D3-14 | D2 |
Gel Size (W x L in cm) | 23 x 14 | 14.4 x 10.2 |
Buffer Capacity (ml) | 800ml | 600ml |
Voltage Requirements (V) | 20-150 | 20-150 |
Time Requirements (min) | 30-60 | 30-60 |
Attachment(s):
File | Last Modified |
---|---|
7007320 - Rev 2 - Horizontal Electrophoresis System, Model D2 and D3-14 - Operating and Maintenance Manual.pdf | December 15, 2021 |