7007325 - Rev 1 - Horizontal Electrophoresis System, Model D4 - Operating and Maintenance Manual
Description:
Operating and maintenance manual
Environment:
Owl Horizontal Agarose Gel Electrophoresis System, model:
- D4
Specifications and recommended running conditions:
Gel size (W x L cm) | 16 x 17 | |
Buffer capacity | 800 | 1 tray |
1600 | 2 trays | |
Voltage requirements (V) | 100V minimum | |
Time requirements | 4½ hours |
Attachment(s):
File | Last Modified |
---|---|
7007325 - Rev 1 - Horizontal Electrophoresis System, Model D4 - Operating and Maintenance Manual.pdf | December 15, 2021 |