7007335 - Rev 1 - Water Cooled, Dual Gel Electrophoresis System, Models P8DS, P9DS and P10DS - Operating and Maintenance Manual
Description:
Operating and maintenance manual
Environment:
Water Cooled, Dual Gel Electrophoresis System, Models:
- P8DS
- P9DS
- P10DS
Specifications:
Unit/Model Number | P8DS | P9DS | P10DS |
Gel size |
10cmW x 10cmL 10cmW x 8cmL |
16cmW x 14cmL 16cmW x 16cmL |
20cmW x 20cmL |
Upper Buffer Chamber Capacity | 170ml | 400ml | 600ml |
Lower Buffer Chamber Capacity | 240ml | 300ml | 800ml |
Total Running Buffer | 250ml | 650ml | 1250ml |
Total Buffer Capacity | 250ml | 650ml | 1250ml |
Current, Constant | 15-35mA/gel | 15-50mA/gel | 15-75mA/gel |
Time Requirements | 30-90 min. | 60-120 min. | 60-180 min. |
Sample Capacity | 20 | 30 | 40 |
Dimensions (cm) H x Wx D | 15 x 17 x 12 | 21 x 24 x 14 | 24 x 30.5 x 16.5 |
Glass Size (cm) Wx L | 10 x 10 | 16 x 14 | 20 x 20 |
Attachment(s):
File | Last Modified |
---|---|
7007335 - Rev 1 - Water Cooled, Dual Gel Electrophoresis System, Models P8DS, P9DS and P10DS - Operating and Maintenance Manual.pdf | January 19, 2022 |