7007341 - Rev 1 - Single Sided Vertical Electrophoresis System, P81 - Operating and Maintenance Manual
Description:
Operating and maintenance manual
Environment:
Single Sided Vertical Electrophoresis System, P81, specifications:
Unit/Model | Number P81 |
Gel size | 10cmW x 10cmL and 10cmW x 8cmL |
Upper Buffer Chamber Capacity | 100ml |
Lower Buffer Chamber Capacity | 50ml |
Total Running Buffer | 150ml |
Total Buffer Capacity | 150ml |
Current, Constant | 15-35mA/gel |
Time Requirements | 30-90 min. |
Sample Capacity | 10 |
Dimensions (cm) H x W x D | 12.7 x 18.4 x 8.3 |
Glass Size (cm)W x L | 10 x 10 |
Attachment(s):
File | Last Modified |
---|---|
7007341 - Rev 1 - Single Sided Vertical Electrophoresis System, P81 - Operating and Maintenance Manual.pdf | December 15, 2021 |