7007351 - Rev 0 - Tank Electroblotting System, Models VEP-2 and VEP-3 - Operating and Maintenance Manual
Description:
Operating and maintenance manual
Environment:
Tank Electroblotting System, models:
- VEP-2
- VEP-3
Specifications and recommended running conditions:
Model VEP-2 | |
Dimensions (Footprint): |
5.125in W x 6.44in H x 7.3in D (13cm W x 16.3cm H x 18.5cm D) |
Buffer required: | 1300ml |
Gel size(maximum): | 10cm x 10cm |
Current requirements: | 200-500mA |
Time required: | 45 minutes to 2 hours |
Model VEP-3 | |
Dimensions (Footprint): | 30cm W x 30cm H x 9cm D |
Buffer required: | 4.0L |
Gel size(maximum): | 20cm x 30cm |
Current requirements: | 1.5 A |
Time required: | 60-120 min. |
Attachment(s):
File | Last Modified |
---|---|
7007351 - Rev 0 - Tank Electroblotting System, Models VEP-2 and VEP-3 - Operating and Maintenance Manual.pdf | December 15, 2021 |