7007359 - Rev 0 - Tank Electroblotting System, Model VEP-2 XM - Installation and Operation Manual
Description:
Installation and operation manual
Environment:
Tank Electroblotting System, Model VEP-2 XM, specifications and recommended running conditions:
Model VEP-2 | |
Dimensions (Footprint): |
5.125in W x 6.44in H x 7.3in D (13cm W x 16.3cm H x 18.5cm D) |
Buffer required: | 1300ml |
Gel size(maximum): | 10cm x 10cm |
Current requirements: | 200-500mA |
Time required: | 45 minutes to 2 hours |
Attachment(s):
File | Last Modified |
---|---|
7007359 - Rev 0 - Tank Electroblotting System, Model VEP-2 XM - Installation and Operation Manual.pdf | December 15, 2021 |