7217023 - Rev 0 - Adjustable Vertical Electrophoresis System, Models ADJ1, ADJ2 and ADJ3 - Operating and Maintenance Manual
Description:
Operating and maintenance manual
Environment:
Adjustable Vertical Electrophoresis System, models:
- ADJ1
- ADJ2
- ADJ3
Specifications:
Unit | ADJ-1 | ADJ-2 | ADJ-3 |
Glass size | 16cm W x 14-40cm L | 20cm W x 20-70 cm L | 35cm W x 20-80cm L |
Overall dimensions | 48cm H x 23cm W x 25cm D | 78cm H x 28cm W x 25cm D | 86cm H x 41 cm W x 25cm D |
Buffer volume | 500ml | 900ml | 1800ml |
Attachment(s):
File | Last Modified |
---|---|
7217023 - Rev 0 - Adjustable Vertical Electrophoresis System, Models ADJ1, ADJ2 and ADJ3 - Operating and Maintenance Manual.pdf | December 15, 2021 |