7217584 - Rev 0 - Electrophoresis Power Supply, EC3000XL - Operating and Maintenance Manual
Description:
Operating and maintenance manual
Environment:
Thermo Scientific Electrophoresis Power Supply, EC3000XL
AC Input Power
- 95-265VAC, 50-60Hz, 500VA Max
Environmental
- Operating temperature: 0-40°C, 0-95% R.H. non-condensing
- Altitude: 2000m
- Overvoltage category II, IEC664
- Pollution degree 2, IEC664
DC Output Power
- 20-3000VDC, 400W Max
- 1-400mA, 400W Max
- Ripple: ±1%
- Drift: ±1%, after 30-minute warm-up
Attachment(s):
File | Last Modified |
---|---|
7217584 - Rev 0 - Electrophoresis Power Supply, EC3000XL - Operating and Maintenance Manual.pdf | December 15, 2021 |