7217692 - Rev 0 - Gel Caster, Models JGC-3, JGC-2, and JGC-4 - Operating and Maintenance Manual
Description:
Operating and maintenance manual. This patented Gel Casting System is a unique method to cast 1 to 4 polyacrylamide gels without taping or special sealing of the gel plates.
Environment:
Gel Caster Models:
- JGC-3
- JGC-2
- JGC-4
Specifications:
Unit/Model Number | JGC-4 | JGC-2 | JGC-3 |
Gel size |
10cmW x 10cmL 10cmW x 8cmL |
16cmW x 14cmL 16cmW x 16cmL |
20cmW x 20cmL |
Footprint (cm) W x D | 13 x 13 | 17 x 18 | 17 x 23 |
Glass Size (cm) W x L |
10cmW x 10cmL 10cmW x 8cmL |
16cmW x 14cmL 16cmW x 16cmL |
20cmW x 20cmL |
Approximate Gel Volume | 6ml | 15ml | 28ml |
Compatible with Unit | P8DS, P81, P82 | P9DS | P10DS |
Attachment(s):
File | Last Modified |
---|---|
7217692 - Rev 0 - Gel Caster, Models JGC-3, JGC-2, and JGC-4 - Operating and Maintenance Manual.pdf | December 15, 2021 |