7217837 - Rev 0 - Multiple Gradient Caster, Models P7-CST, P1-CST and P2-CST - Operating and Maintenance Manual
Description:
Operating and maintenance manual
Environment:
Multiple Gradient Caster, models:
- P7-CST
- P1-CST
- P2-CST
Specifications:
Unit/Model Number | P7-CST | P1-CST | P2-CST |
Gel size |
10cm W x 10cm L 10cm W x 8cm L |
16cm W x 14cm L 16cm W x 16cm L |
20cm W x 20cm L |
Sample Capacity | 20 | 30 | 40 |
Dimensions (cm) W x D x H | 15 x 13 x 13 | 20 x 13 x 18 | 25 x 13 x 23 |
Glass Size(cm)WxL | 10 x 10 | 16 x 14 | 20 x 20 |
Compatible w/ Owl Units | P8DS, P82 | P9DS | P10DS |
Attachment(s):
File | Last Modified |
---|---|
7217837 - Rev 0 - Multiple Gradient Caster, Models P7-CST, P1-CST and P2-CST - Operating and Maintenance Manual.pdf | December 15, 2021 |