7218313 - Rev 0 - Sequencing Gel Casters, Models SGC-1 and SGC-2 - Operating and Maintenance Manual
Description:
Operating and maintenance manual
Environment:
Sequencing Gel Casters, models:
- SGC-1
- SGC-2
SGC-1:
- Gel Size: 20-42cm W x 48cm L
- Footprint: 21.6cm W x 71cm D (fully closed) x 12.2cm H
SGC-2:
- Gel Size: 20-42cm W x 65.5cm L
- Footprint: 21.2cm W x 102cm D (fully closed) x 12.2cm H
Attachment(s):
File | Last Modified |
---|---|
7218313 - Rev 0 - Sequencing Gel Casters, Models SGC-1 and SGC-2 - Operating and Maintenance Manual.pdf | December 15, 2021 |