7219011 Rev 0 - Horizontal System Model A2-OK - Operating and Maintenance Manual
The Horizontal System A2-OK - Operating and Maintenance Manual" provides detailed instructions on the operation and maintenance of the Horizontal System Model A2-OK. It emphasizes the importance of following safety guidelines, including disconnecting the power supply before maintenance and ensuring that only qualified personnel perform internal adjustments. The manual includes specifications, compliance with IEC 1010-1 safety standards, and encourages users to register their warranty online.
Description:
Operating and maintenance manual
Environment:
Horizontal System Model A2-OK, specifications:
| Gel size | 7cmW x 8cmL |
| Running Buffer | Volume 2.3L |
| Footprint | 28cm W x 37.5cm L x 10cm H |
Attachment(s):
| File | Last Modified |
|---|---|
| 7219011 - Rev 0 - Horizontal System, Model A2-OK - Operating and Maintenance Manual.pdf | December 15, 2021 |