Owl EasyCast Horizontal Minigel System and Buffer Puffer Horizontal System - Owner's Manual
The Owl EasyCast Horizontal Minigel System and Buffer Puffer Horizontal System - Owner's Manual provides detailed specifications and recommended running conditions for the models B1A, B1, B2, and B3. It includes information on gel sizes, buffer capacities, time, and voltage requirements. The manual is essential for understanding the operation and maintenance of these electrophoresis systems. For complete details, refer to the owner's manual.
Description:
Owner's manual
Environment:
The Owl EasyCast Horizontal Minigel System, models:
- B1A
- Bl
- B2
The Buffer Puffer Horizontal System, model:
- B3
Specifications and recommended running conditions:
| Model | B1A | B1 | B2 |
| Gel Size (W x L in cm.) | 7x8 | 9x11 | 12x14 |
| Buffer Capacity | 400ML | 600ML | 800ML |
| Time Requirements (minutes) | 30-60 | 45-90 | 60-120 |
| Voltage Requirements (V) | 20-150 | 20-150 | 20-150 |
Model B3
Gel Size W x L in cm. 12x14
Buffer Caacit 100ML
Time Reuirements min. 60-120
Voltaoe Reuirements V 20-150
Attachment(s):
| File | Last Modified |
|---|---|
| Owl EasyCast Horizontal Minigel System and Buffer Puffer Horizontal System - Owner's Manual.pdf | January 19, 2022 |