The Opossum Horizontal System, Model A2-0K - Owner's Manual
Description:
Owner's Manual
Environment:
Owl Separation Systems, The Opossum Horizontal Multiple Gel, Model A2-OK system
Specifications:
Gel size | 7cmW x 8cmL (x6) |
Running Buffer Volume | 2.3L |
Footprint | 28cmW x 37.5cmL x 10cmH |
Attachment(s):
File | Last Modified |
---|---|
The Opossum Horizontal System, Model A2-0K - Owner's Manual.pdf | January 19, 2022 |