103770 - Fisher Scientific Isotemp Programmable Muffle Furnace_650-750 Series - Instruction Manual
The Fisher Scientific Isotemp Programmable Muffle Furnace 650-750 Series Instruction Manual details installation, operation, and safety for models 14, 14A, 58, and 126. Key points include wearing safety gear, selecting a proper installation location, and removing packing materials before use. The furnace features PID microprocessor temperature control up to 1125°C, with fast heat-up and cool-down. The model 750 can store four 24-step programs, while the model 650 stores one.
Description:
Instruction manual
Environment:
Fisher Scientific Isotemp® Programmable Muffle Furnace, 650-750 Series:
| Model | |
| Model 14 | 120 V, 50/60 Hz |
| Model 14A | 208/240V, 50/60 Hz |
| Model 58 | 208/240 V, 50/60 Hz |
| Model 126 | 208/240 V, 50/60 Hz |
Attachment(s):
| File | Last Modified |
|---|---|
| 103770 - Fisher Scientific Isotemp Programmable Muffle Furnace, 650-750 Series - Instruction Manual.pdf | December 16, 2021 |