2116 Temperature Controller - Temperature Offset Calibration
To calibrate the temperature offset for the 2116 Temperature Controller, you need an N.I.S.T. traceable type K thermocouple with a digital pyrometer. Allow the furnace to stabilize at the set temperature. Record temperatures from both the furnace controller and the pyrometer. Calculate the offset by subtracting the furnace controller temperature from the pyrometer reading. Enter this offset into the controller and verify the calibration by checking if the temperatures match closely after stabili
Description:
Temperature offset calibration procedure
Environment:
2116 Temperature Controller
Attachment(s):
| File | Last Modified |
|---|---|
| 2116 Temperature Controller - Temperature Offset Calibration.pdf | December 17, 2021 |