Thermo Fisher Scientific
330985H01 - Thermo Scientific Lindberg/Blue M Moldatherm 1100C Box Furnace - Installation and Operational Manual
Description: 
Installation and operational manual
Environment:
Thermo Scientific Lindberg/Blue M Moldatherm 1100°C Box Furnace, models:
Model  
BF51748A-1 120 VAC, 50/60 Hz, 1800 W, 1 Phase, 15 A
BF51748COMA-1# 120 VAC, 50/60 Hz, 1800 W, 1 Phase, 15 A
BF51748C-1 208/240 VAC, 50/60 Hz, 1800 W, 1 Phase, 7.5 A
BF51766A-1 120 VAC, 50/ 60 Hz, 1800 W, 1 Phase, 15 A
BF51766COMA-1# 120 VAC, 50/ 60 Hz, 1800 W, 1 Phase, 15 A
BF51766C-1 208/240 VAC, 50/60Hz, 1800 W, 1 Phase, 7.5 A
BF51794C-1 208/240 V, 50/60 Hz, 3500 W, 14.6 A
BF51728C-1 208/240 V, 50/60 Hz, 5600 W, 23.3 A
BF51728RHDC-1 208/240 V, 50/60 Hz, 5600 W, 23.3 A
BF51848A-1 120 VAC, 50/60 Hz, 1800 W, 1 Phase, 15 A
BF51848COMA-1# 120 VAC, 50/60 Hz, 1800 W, 1 Phase, 15 A
BF51848C-1 208/240 VAC, 50/60 Hz, 1800 W, 1 Phase, 7.5 A
BF51848COMC-1 208/240 VAC, 50/60 Hz, 1800 W, 1 Phase, 7.5 A
BF51866A-1 120 VAC, 50/ 60 Hz, 1800 W, 1 Phase, 15 A
BF51866COMA-1# 120 VAC, 50/ 60 Hz, 1800 W, 1 Phase, 15 A
BF51866C-1 208/240 VAC, 50/60Hz, 1800 W, 1 Phase, 7.5 A
BF51894C-1 208/240 V, 50/60 Hz, 3500 W, 14.6 A
BF51828C-1 208/240 V, 50/60 Hz, 5600 W, 23.3 A
BF51828RHDC-1 208/240 V, 50/60 Hz, 5600 W, 23.3 A
#Unit includes male and female RS 485 Digital Communications DB-9 ports.