39129H01-RC - Lindberg/Blue Moldatherm Box Furnaces, BF51600, BF51700, and BF51800 Series - Installation and Operation Manual
Description:
Installation and operation manual
Environment:
Lindberg/Blue Moldatherm, BF51600, BF51700, and BF51800 Series Moldatherm Box Furnaces, models:
Model | Watts | Voltage | |
BF51648Aa | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
BF51648Ca | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
BF51748Aa | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
BF51748Ca | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
BF51848Aa | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
BF51848Ca | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
BF51666Aa | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
BF51666Ca | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
BF51766Aa | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
BF51766Ca | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
BF51866Aa | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
BF51866Ca | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
BF51694Cb | 3500 | 208/240 VAC 50/60 HZ, 1 Phase | |
BF51794Cb | 3500 | 208/240 VAC 50/60 HZ, 1 Phase | |
BF51894Cb | 3500 | 208/240 VAC 50/60 HZ, 1 Phase | |
BF51628C | 5600 | 208/240 VAC 50/60 HZ, 1 Phase | |
BF51728C | 5600 | 208/240 VAC 50/60 HZ, 1 Phase | |
BF51828C | 5600 | 208/240 VAC 50/60 HZ, 1 Phase |
aVoltage Code: A - 120V Model; C - 208/240V Model. “A" models are equipped with power cord and plug for 120 VAC operation. “C" models are equipped with power cord and plug for 208/240 VAC operation.
bEquipped with power cord and plug for 240 VAC operation. q
Attachment(s):
File | Last Modified |
---|---|
39129H01-RC - Lindberg_Blue Moldatherm Box Furnaces, BF51600, BF51700, and BF51800 Series - Installation and Operation Manual.pdf | December 17, 2021 |