330985H01 Rev A and C - Thermo Scientific Lindberg Blue M Moldatherm 1100C Box Furnace - Installation and Operational Manual
The installation and operational manuals for the Thermo Scientific Lindberg/Blue M Moldatherm 1100°C Box Furnace, revisions A and C (document 330985H01), cover models BF51728, BF51748, BF51766, BF51794, BF51828, BF51848, BF51866, and BF51894. They include safety notes, installation instructions, operational guidelines, and maintenance procedures. Rev A was published in April 2020, and Rev C in October 2020.
Description:
Installation and operational manual
Environment:
Thermo Scientific Lindberg/Blue M Moldatherm 1100°C Box Furnace, models:
| Model | |
| BF51748A-1 | 120 VAC, 50/60 Hz, 1800 W, 1 Phase, 15 A |
| BF51748COMA-1# | 120 VAC, 50/60 Hz, 1800 W, 1 Phase, 15 A |
| BF51748C-1 | 208/240 VAC, 50/60 Hz, 1800 W, 1 Phase, 7.5 A |
| BF51766A-1 | 120 VAC, 50/ 60 Hz, 1800 W, 1 Phase, 15 A |
| BF51766COMA-1# | 120 VAC, 50/ 60 Hz, 1800 W, 1 Phase, 15 A |
| BF51766C-1 | 208/240 VAC, 50/60Hz, 1800 W, 1 Phase, 7.5 A |
| BF51794C-1 | 208/240 V, 50/60 Hz, 3500 W, 14.6 A |
| BF51728C-1 | 208/240 V, 50/60 Hz, 5600 W, 23.3 A |
| BF51728RHDC-1 | 208/240 V, 50/60 Hz, 5600 W, 23.3 A |
| BF51848A-1 | 120 VAC, 50/60 Hz, 1800 W, 1 Phase, 15 A |
| BF51848COMA-1# | 120 VAC, 50/60 Hz, 1800 W, 1 Phase, 15 A |
| BF51848C-1 | 208/240 VAC, 50/60 Hz, 1800 W, 1 Phase, 7.5 A |
| BF51848COMC-1 | 208/240 VAC, 50/60 Hz, 1800 W, 1 Phase, 7.5 A |
| BF51866A-1 | 120 VAC, 50/ 60 Hz, 1800 W, 1 Phase, 15 A |
| BF51866COMA-1# | 120 VAC, 50/ 60 Hz, 1800 W, 1 Phase, 15 A |
| BF51866C-1 | 208/240 VAC, 50/60Hz, 1800 W, 1 Phase, 7.5 A |
| BF51894C-1 | 208/240 V, 50/60 Hz, 3500 W, 14.6 A |
| BF51828C-1 | 208/240 V, 50/60 Hz, 5600 W, 23.3 A |
| BF51828RHDC-1 | 208/240 V, 50/60 Hz, 5600 W, 23.3 A |
#Unit includes male and female RS 485 Digital Communications DB-9 ports.