LT1060X1 - Barnstead Thermolyne Type F6000 Furnace Series 1060 - Operation Manual and Parts List
The document LT1060X1 for the Barnstead Thermolyne Type F6000 Furnace Series 1060 includes the operation manual and parts list. It covers various models such as F6010, F6020C, and F6030CM, detailing their voltage, control types, display settings, and RS232 communication capabilities. The manual provides essential operating and safety information, which must be thoroughly read and understood before using the equipment.
Description:
Operation manual and parts list
Environment:
Barnstead Thermolyne, Type F6000 Furnace, Series 1060, model numbers:
| Model | Voltage | Control | Display (°C and °F) | RS232 Comm. |
| F6010 | 220-240 | Basic Automatic | X (°C only) | |
| F6010-33 | 220-240 | Basic Automatic | X (°C only) | |
| F6018 | 208 | Basic Automatic | X (°C only) | |
| F6020C | 220-240 | Automatic | X | |
| F6020C-33 | 220-240 | Automatic | X | |
| F6020C-33-60 | 220-240 | Automatic | X | |
| F6020C-33-60-80 | 220-240 | Programmable | X | |
| F6020C-33-80 | 220-240 | Programmable | X | |
| F6020C-60 | 220-240 | Automatic | X | |
| F6020C-60-80 | 220-240 | Programmable | X | |
| F6020C-80 | 220-240 | Programmable | X | |
| F6028C | 208 | Automatic | X | |
| F6028C-60 | 208 | Automatic | X | |
| F6028C-60-80 | 208 | Programmable | X | |
| F6028C-80 | 208 | Programmable | X | |
| F6030CM | 220-240 | Multi-Programmable | X | X |
| F6030CM-33 | 220-240 | Multi-Programmable | X | X |
| F6030CM-33-60 | 220-240 | Multi-Programmable | X | X |
| F6030CM-60 | 220-240 | Multi-Programmable | X | X |
| F6038CM | 208 | Multi-Programmable | X | X |
| F6038CM-60 | 208 | Multi-Programmable | X | X |
(-60 Models) - Ashing furnace
Attachment(s):
| File | Last Modified |
|---|---|
| LT1060X1 - Barnstead Thermolyne, Type F6000 Furnace, Series 1060 - Operation Manual and Parts List.pdf | December 16, 2021 |