Lindberg Blue M 1100C Rated Chambers - Furnace Exhaust Port Ventilation Connection
This document provides a troubleshooting guide for the ventilation connection of the furnace exhaust port in Lindberg Blue M 1100°C rated chambers. Proper ventilation is crucial for removing vapors and gases during high-temperature operations. The exhaust port is 1 inch in diameter and located through the top wall. Use an exhaust hood or metal tube/pipe with proper spacing to minimize the chimney effect. Consult an HVAC technician for optimal setup.
Description:
Troubleshooting guide for furnace exhaust port ventilation connection
Environment:
Lindberg Blue M 1100C Rated Chambers
Attachment(s):
| File | Last Modified |
|---|---|
| Lindberg Blue M 1100C Rated Chambers - Furnace Exhaust Port Ventilation Connection.pdf | December 17, 2021 |