Lindberg Blue M 1700C Tube Furnaces - Vestibule and Sleeve Applications Troubleshooting Guide
The Lindberg Blue M 1700C Tube Furnaces - Vestibule and Sleeve Applications Troubleshooting Guide provides detailed information on using and troubleshooting vestibules and sleeves in these high-temperature furnaces. Vestibules support the Process tube with insulation, while sleeves insulate the Process tube. Proper positioning of sleeves against vestibules ensures smooth temperature transitions and prevents fractures. The guide includes steps for diagnosing and resolving common issues.
Description:
Troubleshooting guide
Environment:
Lindberg Blue M 1700C Tube Furnaces Vestibule and Sleeve Applications
Attachment(s):
| File | Last Modified |
|---|---|
| Lindberg Blue M 1700C Tube Furnaces - Vestibule and Sleeve Applications Troubleshooting Guide.pdf | December 17, 2021 |