Lindberg Blue M Ashing Furnaces - Guidelines for Ashing Applications
For Lindberg Blue M Ashing Furnaces, it is crucial to prevent carbon residue buildup by regularly operating the furnace at temperatures above 900°C for one hour. This practice helps remove carbon deposits, which can cause electrical short circuits and damage the heating elements. Adhering to the manufacturer's guidelines and performing regular maintenance ensures optimal furnace performance.
Description:
Troubleshooting guide and guidelines for Ashing applications
Environment:
Lindberg Blue M Ashing Furnaces
Attachment(s):
| File | Last Modified |
|---|---|
| Lindberg Blue M Ashing Furnaces - Guidelines for Ashing Applications.pdf | December 17, 2021 |