Temperature Control Autotuning Procedure
To autotune the 3216 temperature control furnace, load the chamber with materials similar to a normal load and close the door. Set the setpoint temperature as required. Press the SCROLL button until "A.TUNE" is displayed, then select "ON" using the UP or DOWN button. Press the PAGE button to return to the HOME display, where "TUNE" will flash during the tuning process. The process completes when the regular temperature display returns.
Issue
3216 temperature control furnace autotuning procedure
Environment
3216 Eurotherm
Resolution
Refer to the attached document for the procedure
Attachment(s)
| File | Last Modified |
|---|---|
| Temperature Control Autotuning Procedure.pdf | January 13, 2022 |