Thermocouple Errors E 001 and E 002
Thermocouple Errors E 001 and E 002 in the NCAT Asphalt Furnace indicate issues with the thermocouple sensors in the Chamber and Filter areas, respectively. Both errors suggest the thermocouple is open, failed, or disconnected. Replacing the respective thermocouple usually resolves the issue. If not, verify wiring connections and measure continuity.
Issue
Thermocouple Errors E 001 and E 002
Environment
NCAT Asphalt Furnace
Resolution
Refer to the attached document for information.
Attachment(s)
| File | Last Modified |
|---|---|
| Thermocouple Errors E 001 and E 002.pdf | January 13, 2022 |