304069H01 - Lindberg Blue M MO 1420-1450 Series Mechanical Convection Ovens - Installation and Operation Manual
The installation and operation manual for the Lindberg/Blue M MO 1420-1450 Series Mechanical Convection Ovens covers models MO1420A, MO1430A, MO1440A, MO1440C, MO1450A, MO1450C, MO1420SA, MO1440SA, MO1440SC, and MO1450SA. It includes details on installation, operation, and maintenance procedures. The manual also covers safety considerations, troubleshooting steps, and wiring diagrams. The ovens feature a microprocessor-based temperature controller and an internal blower for temperature uniformit
Description:
Installation and operation manual
Environment:
Lindberg/Blue M MO 1420-1450 Series Mechanical Convection Ovens, models:
| Model | Voltage |
| MO1420A | 120 V, 50/60 Hz, Single Phase |
| MO1430A | 120 V, 50/60 Hz, Single Phase |
| MO1440A | 120 V, 50/60 Hz, Single Phase |
| MO1440C | 240 V, 50/60 Hz, Single Phase |
| MO1450A | 120 V, 50/60 Hz, Single Phase |
| MO1450C | 240 V, 50/60 Hz, Single Phase |
| MO1420SA | 120 V, 50/60 Hz, Single Phase |
| MO1440SA | 120 V, 50/60 Hz, Single Phase |
| MO1440SC | 240 V, 50/60 Hz, Single Phase |
| M01450SA | 120 V, 50/60 Hz, Single Phase |
Attachment(s):
| File | Last Modified |
|---|---|
| 304069H01 - Lindberg_Blue M MO 1420-1450 Series Mechanical Convection Ovens - Installation and Operation Manual.pdf | December 20, 2021 |