3177873 - Rev E - NAPCO Vacuum Oven, Model 6508/6509 (5861) - User Manual
The user manual for the NAPCO Vacuum Oven, Model 6508/6509 (5861), includes sections on unpacking, installation, specifications, controls, and temperature adjustment. It provides instructions for positioning and leveling the oven, installing shelves, and maintaining safety clearances. The manual details the oven's technical specifications, control functions, and temperature setting procedures.
Description:
User manual
Environment:
NAPCO® Vacuum Oven, model 6508/6509 (5861). Catalog numbers 3166547 and 3166548.
| Standard electrical service: | 1500 Watts, 120 VAC, 50/60 Hz, 12.5 Amps |
| Optional electrical service: | 1500 Watts, 240 VAC, 50/60 Hz, 6.3 Amps |
Attachment(s):
| File | Last Modified |
|---|---|
| 3177873 - Rev E - NAPCO Vacuum Oven, Model 6508_6509 (5861) - User Manual.pdf | December 20, 2021 |