3177885 - Rev J - Thelco Oven, Model 6540 and 6550 Series - Operating and Maintenance Manual
The manual for Thelco Oven models 6540 and 6550 series provides detailed operational and maintenance instructions. It covers various models, specifying voltage requirements, temperature control ranges, and environmental conditions for optimal performance. The document includes safety precautions, component descriptions, and technical specifications for both mechanical and gravity convection models.
Description:
Operating and maintenance manual
Environment:
Thelco Oven, model 6540 and 6550 series, model numbers:
| Current Model # | Previous Model # | Voltage |
| 6540 | 70 | 120V |
| 6541 | 70 | 230V |
| 6545 | 70D | 120V |
| 6546 | 70D | 230V |
| 6550 | 70M | 120V |
| 6551 | 70M | 230V |
| 6555 | 70DM | 120V |
| 6556 | 70DM | 230V |
| 6542 | 130 | 120V |
| 6543 | 130 | 230V |
| 6547 | 130D | 120V |
| 6548 | 130D | 230V |
| 6552 | 130M | 120V |
| 6553 | 130M | 230V |
| 6557 | 130DM | 120V |
| 6558 | 130DM | 230V |
| 6544 | 160 | 230V |
| 6549 | 160D | 230V |
| 6554 | 160M | 230V |
| 6559 | 160DM | 230V |
Catalog numbers:
| Specification | Model 6540/6541 | Model 6542/6543 | Model 6544 | Model 6545/6546 | Model 6547/6548 | Model 6549 |
| Electrical 50/60 Hz 115V | 1200W 10.4A | 1700W 14.8A | N/A N/A | 1200W 10.4A | 1700W 14.8A | N/A N/A |
| Electrical 50/60 Hz 230V | 1200W 5.2A | 1700W 7.4A | 2000W 8.7A | 1200W 5.2A | 1700W 7.4A | 2000W 8.7A |
| Catalog Number 115V | 3166782 | 3166784 | -- | 3166787 | 3166789 | -- |
| Catalog Number 230V | 3166783 | 3166785 | 3166786 | 3166788 | 3166790 | 3166791 |
Attachment(s):
| File | Last Modified |
|---|---|
| 3177885 - Rev J - Thelco Oven, Model 6540 and 6550 Series - Operating and Maintenance Manual.pdf | December 20, 2021 |