50125548 D - Heratherm General Protocol Heating and Drying Ovens - Operating Instructions
The Thermo Scientific Heratherm General Protocol Heating and Drying Ovens are designed for laboratory heating applications with precision temperature control, suitable for heat treating samples or materials at temperatures between 50 °C and 250 °C. The operating instructions emphasize safety, correct and incorrect uses, and compliance with international standards. Only trained and authorized personnel should operate these ovens, and the instructions must be fully read and understood before use.
Description:
Operating Instructions
Environment:
Thermo Scientific Heratherm General Protocol Heating and Drying Ovens, models:
- Heratherm OGS60
- Heratherm OGS100
- Heratherm OGS180
- Heratherm OGS400
- Heratherm OGS750
- Heratherm OMS60
- Heratherm OMS100
- Heratherm OMS180
- 51028112
- 51028872
- 51028874
- 51029327
- 51029339
- 51028121
- 51028122
- 51028873
- 51028875
- 51028123
Attachment(s):
| File | Last Modified |
|---|---|
| 50125548 D - Heratherm General Protocol Heating and Drying Ovens - Operating Instructions.pdf | December 21, 2021 |