50141333_B_250914 - Thermo Scientific Heratherm - Main Board Kit Assembly Instructions
To assemble the main board kit for the Thermo Scientific Heratherm ovens and incubators, ensure the unit is disconnected from power and follow ESD guidelines. Disconnect all plugs, unscrew the electrical compartment, and replace the main board, reconnecting plugs as previously arranged. The socket locations for heaters and other components are detailed within the assembly instructions.
Description:
Main board kit assembly instructions
Environment:
Thermo Scientific Heratherm, IGS, IMH, OMS, OMH, OMH-S, OGS, OGH, OGH-S and IMH-S ovens and incubators.
Attachment(s):
| File | Last Modified |
|---|---|
| 50141333_B_250914 - Thermo Scientific Heratherm - Main Board Kit Assembly Instructions.pdf | December 20, 2021 |