50141691_A_021014 - Heratherm - Mainboard Kit Assembly Instructions
The Thermo Scientific Heratherm Mainboard Kit includes safety instructions and assembly guidance for floor models, 3-phase units. Key steps involve disconnecting the power and removing the electronics cover, replacing the mainboard according to the wiring diagram, and ensuring all connections are secure and correctly phased. Required tools include a Torx 20x100 screwdriver, and ESD protection is recommended.
Description:
Mainboard kit
Environment:
Main board kit floor 3-Phase
Attachment(s):
| File | Last Modified |
|---|---|
| 50141691_A_021014 - Heratherm - Mainboard Kit Assembly Instructions.pdf | December 20, 2021 |