7000664 - Rev 4 - Gravity Convection Ovens - Installation and Operation Manual
The manual "7000664 - Rev 4 - Gravity Convection Ovens" covers installation, operation, and maintenance for Fisherbrand Isotemp Gravity Convection Ovens, specifically model 3511FSQ (664). It includes safety information, installation requirements, start-up procedures, and maintenance guidelines. The ovens are designed for various laboratory applications, with a temperature range from ambient +5°C to 210°C, controlled by a bimetallic thermostat.
Description:
Installation and operation manual
Environment:
Gravity Convection Ovens, model:
| Model Number | Size | Voltage |
| 3511FSQ (664) | 1.7 cu ft | 120V |
Attachment(s):
| File | Last Modified |
|---|---|
| 7000664 - Rev 4 - Gravity Convection Ovens - Installation and Operation Manual.pdf | December 21, 2021 |