7026240 - Rev 0 - Shake n Stack Hybridisation Oven - Operating and Maintenance Manual
The Shake 'n Stack Hybridisation Oven Operating and Maintenance Manual (7026240 - Rev 0) covers models 6240, 6241, 6242, 6243, 6244, and 6245. It includes detailed instructions for installation, operation, and maintenance. This manual emphasizes safety precautions, such as handling electrostatic discharge and hot surfaces, and ensuring activities involving electrical hazards are performed by qualified personnel. The manual also covers the control panel operations and speed adjustments for the ro
Description:
Operating and maintenance manual
Environment:
Shake ‘n Stack Hybridisation Oven, models:
| Models | Voltage | Includes |
| 6240 | 220 | Shaking platform, drip tray, rotisserie |
| 6241 | 110 | Shaking platform, drip tray, rotisserie |
| 6242 | 220 | Drip tray, rotisserie |
| 6243 | 110 | Drip tray, rotisserie |
| 6244 | 110 | Drip tray |
| 6245 | 110 | Shaking platform, drip tray |
Attachment(s):
| File | Last Modified |
|---|---|
| 7026240 - Rev 0 - Shake n Stack Hybridisation Oven - Operating and Maintenance Manual.pdf | December 21, 2021 |