7177897 - Rev 0 - Freas Micro-Controlled Ovens Models 605_605P_625_and 645 - Installation Guide
This installation guide provides detailed instructions for installing Freas Micro-Controlled Ovens, specifically models 605, 605P, 625, and 645. It includes environmental details such as voltage, frequency, phase, total watts, total amps, blower motor specifications, and maximum rated temperature. The guide emphasizes the importance of following the instructions to avoid damage, injury, and performance issues. It also includes safety precautions, warranty registration information, and notes abou
Description:
Installation guide
Environment:
Freas Micro-Controlled Ovens, models 605, 605P, 625, and 645:
| Model No. | Volts | Hz | Phase | Total Watts | Total Amps | Blower Motor | Maximum Rated Temp (in °C) |
| 6050/6051 | 120 | 50/60 | 1 | 2500 | 20.8 | 1/6 H.P. | 325 |
| 6052/6053 | 230 | 50/60 | 1 | 2500 | 11 | 1/6 H.P. | 325 |
| 6054 | 230 | 50/60 | 1 | 3700 | 16 | 1/6 H.P. | 325 |
| 6056 | 230 | 50/60 | 1 | 4800 | 21 | 1/6 H.P. | 325 |
Attachment(s):
| File | Last Modified |
|---|---|
| 7177897 - Rev 0 - Freas Micro-Controlled Ovens Models 605, 605P, 625, and 645 - Installation Guide.pdf | December 20, 2021 |