Thermo Fisher Scientific
7177897 - Rev 1 - Precision High Performance Ovens Models 605, 605P, 625, and 645 - Installation Guide
Description: 
This installation guide contains the instructions for Precision High Performance Ovens Models 605, 605P, 625, and 645.
Environment:
Model No.     Volts    Hz    Phase    Total Watts    Total Amps    Blower Motor    Maximum Rated Temp in C
  • 6050               120    50/60     1             2500             20.8             1/6 H.P.                 325
  • 6052
  • 6051               230    50/60     1             2500             11                 1/6 H.P.                325
  • 6053
  • 6054              230    50/60     1             3700             16                 1/6 H.P.                325
  • 6056              230    50/60     1             4800             21                 1/6 H.P.                325