7990652 - Rev 2 - Precision Compact Gravity Convection Oven - Installation Guide
The installation guide for the Precision Compact Gravity Convection Oven covers models PR305225G, PR305220G, and PR305220GCN. It includes detailed instructions for inspecting the shipping carton, setting the oven location, meeting electrical requirements, installing shelves, connecting power, and installing a thermometer. The guide emphasizes the importance of using properly grounded outlets and following local electrical codes. It also highlights the need to inspect for shipping damage promptly
Description:
Installation guide
Environment:
Precision Compact Gravity Convection Oven, models:
| Model: | Power Requirements: |
| PR305225G | 120 Volts, 50/60 Hz, 6.7 Amps, 800 Watts |
| PR305220G | 240 Volts, 50/60 Hz, 3.3 Amps, 800 Watts |
| PR305220GCN | 240 Volts, 50/60 Hz, 3.3 Amps, 800 Watts |
Attachment(s):
| File | Last Modified |
|---|---|
| 7990652 - Rev 2 - Precision Compact Gravity Convection Oven - Installation Guide.pdf | December 20, 2021 |