7990665 - Rev 2 - Precision Compact Ovens with Mechanical Convection - Installation Guide
This installation guide covers the setup of Precision Compact Ovens with Mechanical Convection, specifically models PR305225M, PR305220M, and PR305220MCN. It includes step-by-step instructions to ensure proper installation and optimal performance. The guide addresses all necessary environmental conditions and provides detailed procedures for safely installing and configuring the ovens in a laboratory setting.
Description:
Installation guide
Environment:
Precision Compact Ovens with Mechanical Convection, models:
| Model: | Power Requirements: |
| PR305225M | 120 Volts, 50/60 Hz, 10 Amps, 1200 Watts |
| PR305220M | 240 Volts, 50/60 Hz, 5 Amps, 1200 Watts |
| PR305220MCN | 240 Volts, 50/60 Hz, 5 Amps, 1200 Watts |
Attachment(s):
| File | Last Modified |
|---|---|
| 7990665 - Rev 2 - Precision Compact Ovens with Mechanical Convection - Installation Guide.pdf | December 20, 2021 |