9-056-602-00 - Rev E - Lab-Line Vacuum Ovens - Operation Manual and Parts List
The "9-056-602-00 - Rev E - Lab-Line Vacuum Ovens - Operation Manual and Parts List" provides comprehensive guidance for operating and maintaining Lab-Line Vacuum Ovens, including models such as 3608, 3608-1, 3608-1ACE, 3608-1CE, 3608-1CCE, 3608-1C/PCE, 3608-5, 3608-5X, 3608X, 3608-6, 3608-6CE, 3608-6CCE, 3618, 3618FEP, 3618-1, 3618-1CE, 3618-1CCE, 3618-5, and 3618-6CE. Key sections include introduction, description, specifications, installation, features, operation, maintenance, troubleshooting
Description:
Operation manual and parts list
Environment:
Lab-Line Vacuum Ovens, models:
| Models | Power Requirements |
| 3608 | 120 VAC, 50/60 Hz, 5.0 Amps, 600 Watts |
| 3608-5 | 120 VAC, 50/60 Hz, 5.0 Amps, 600 Watts |
| 3608-1 | 240 VAC, 50/60 Hz, 2.5 Amps, 600 Watts |
| 3608-6 | 240 VAC, 50/60 Hz, 2.5 Amps, 600 Watts |
| 3618 | 120 VAC, 50/60 Hz, 13.3 Amps, 1600 Watts |
| 3618-5 | 120 VAC, 50/60 Hz, 13.3 Amps, 1600 Watts |
| 3618-1 | 240 VAC, 50/60 HZ, 6.7 Amps, 1600 Watts |
| 3618-6 | 240 VAC, 50/60 HZ, 6.7 Amps, 1600 Watts |
| 3618FEP | (Same as Model 3618 except interior of unit is Teflon® coated) |
Attachment(s):
| File | Last Modified |
|---|---|
| 9-056-602-00 - Rev E - Lab-Line Vacuum Ovens - Operation Manual and Parts List.pdf | December 20, 2021 |