LT1476X7 - Rev 0 - Class 100 Clean Room Ovens - Operating Manual and Parts List
The "LT1476X7 - Rev 0 - Class 100 Clean Room Ovens - Operating Manual and Parts List" details the operation and maintenance of Class 100 Clean Room Ovens. Models include various sizes and voltages, some with HEPA filters. Features include a programmer-controller, mechanical air circulation, and dual overheating protection systems. Safety instructions emphasize the importance of following manual guidelines and having qualified personnel perform maintenance. The manual also covers door gasket repa
Description:
Operating manual and parts list
Environment:
Class "100" Clean Room Ovens, models:
| Models | Size | Voltage | With HEPA Filter |
| 3490M (6879) | 2.6 cu ft | 120V | |
| 3490M-1 (6880) | 2.6 cu ft | 240V | |
| 3492M (6881) | 2.6 cu ft | 120V | |
| 3492M-1 (6882) | 2.6 cu ft | 240V | |
| 3494M-1 (6883) | 2.6 cu ft | 240V | |
| 3495M-1 (6884) | 3.6 cu ft | 240V | |
| 3496M-1 (6885) | 2.8 cu ft | 240V | X |
| 3497M-1 (6886) | 2.8 cu ft | 240V | X |
| 3498M-1 (6887) | 4.6 cu ft | 240V | |
| 3499M-1 (6888) | 4.6 cu ft | 240V |
Attachment(s):
| File | Last Modified |
|---|---|
| LT1476X7 - Rev 0 - Class 100 Clean Room Ovens - Operating Manual and Parts List.pdf | December 21, 2021 |