LT1479X5 (7000662) - Rev 3 - StableTemp Economical Gravity Convection Ovens - User Manual
The user manual for Cole-Parmer StableTemp Economical Gravity Convection Ovens covers models 52120-03 (662) and 52120-02 (663). It includes detailed instructions for installation, operation, and maintenance. The ovens are designed for drying, sterilization, and baking applications with a temperature range from ambient +5°C to 210°C. Safety precautions emphasize the importance of qualified personnel for adjustments and maintenance, and using protective equipment to prevent injuries.
Description:
User manual
Environment:
Cole-Parmer StableTemp Economical Gravity Convection Ovens, models:
| Models | Size | Voltage | |
| 52120-03 (662) | 3511-1C/PQ | 1.7 cu ft | 240V |
| 52120-02 (663) | 3511C/PQ | 1.7 cu ft | 120V |
Attachment(s):
| File | Last Modified |
|---|---|
| LT1479X5 (7000662) - Rev 3 - StableTemp Economical Gravity Convection Ovens - User Manual.pdf | December 21, 2021 |