LT2042X1 - Isotemp Premium Ovens 700 Series - Operation Manual and Parts List
The "LT2042X1 - Isotemp Premium Ovens 700 Series - Operation Manual and Parts List" provides detailed information on the operation, maintenance, and parts of the Isotemp Premium Ovens 700 Series, including Gravity and Forced Air Ovens. It covers models OV700F, OV700G, OV701F, OV701G, OV702F, and OV702G. Key topics include specifications, installation, maintenance, troubleshooting, and a list of replacement parts. The document also includes electrical requirements, temperature ranges, and control
Description:
Operation manual and parts list
Environment:
Isotemp® Premium Ovens 700 Series, Gravity Ovens and Forced Air Ovens, models:
| OV700F ovens | |
| 13-247-725F | 120 V, 11.5A, 1380W, 60 Hz |
| 13-247-726F | 240 V, 5.8A, 1392W, 50/60 Hz |
| OV700G ovens | |
| 13-247-725G | 120 V, 11A, 1320W, 60Hz |
| 13-247-726G | 240 V, 5.5A, 1320W, 50/60Hz |
| OV701F ovens | |
| 13-247-737F | 120 V, 15.5A, 1860W, 60 Hz |
| 13-247-738F | 240 V, 7.8A, 1870W, 50/60 Hz |
| OV701G ovens | |
| 13-247-737G | 120 V, 15A, 1800W, 60Hz |
| 13-247-738G | 240 V, 7.5A, 1800W, 50/60Hz |
| OV702F ovens | |
| 13-247-750F | 120 V, 15.5A, 1860W, 60 Hz |
| 13-247-751F | 240 V, 7.8A, 1872W, 50/60 Hz |
| OV702G ovens | |
| 13-247-750G | 120 V, 15A, 1800W, 60Hz |
| 13-247-751G | 240 V, 7.5A, 1800W, 50/60Hz |
Attachment(s):
| File | Last Modified |
|---|---|
| LT2042X1 - Isotemp Premium Ovens 700 Series - Operation Manual and Parts List.pdf | December 20, 2021 |