LT2148X1 (7006916) - Rev 3 - Isotemp Premium Ovens 700 Series - Operation Manual and Parts List
The LT2148X1 (7006916) - Rev 3 manual provides detailed operational and maintenance instructions, along with parts lists for the Isotemp Premium Ovens 700 Series. It covers models 13247725F/G, 13247726F/G, 13247737F/G, 13247738F/G, 13247750F/G, and 13247751F/G. These ovens come in three sizes (Small, Medium, Large) and offer both forced air and gravity configurations, with temperature ranges from 50°C to 275°C.
Description:
Operation manual and parts list
Environment:
Isotemp® Premium Ovens 700 Series, models:
| Model | Size | Voltage | Forced Air/Gravity |
| 13247725F (6916) | Small (2.5 cu ft) | 120V | Forced Air |
| 13247725G (6917) | Small (2.5 cu ft) | 120V | Gravity |
| 13247726F (6918) | Small (2.5 cu ft) | 240V | Forced Air |
| 13247726G (6919) | Small (2.5 cu ft) | 240V | Gravity |
| 13247737F (6921) | Medium (3.8 cu ft) | 120V | Forced Air |
| 13247737G (6922) | Medium (3.8 cu ft) | 120V | Gravity |
| 13247738F (6923) | Medium (3.8 cu ft) | 240V | Forced Air |
| 13247738G (6924) | Medium (3.8 cu ft) | 240V | Gravity |
| 13247750F (6925) | Large (5 cu ft) | 120V | Forced Air |
| 13247750G (6926) | Large (5 cu ft) | 120V | Gravity |
| 13247751F (6927) | Large (5 cu ft) | 240V | Forced Air |
| 13247751G (6928) | Large (5 cu ft) | 240V | Gravity |
Attachment(s):
| File | Last Modified |
|---|---|
| LT2148X1 (7006916) - Rev 3 - Isotemp Premium Ovens 700 Series - Operation Manual and Parts List.pdf | December 21, 2021 |