LT2148X2 - Rev 1 - Precision Premium Ovens, 3050 Series - Operating Manual and Parts List
The "LT2148X2 - Rev 1 - Precision Premium Ovens, 3050 Series - Operating Manual and Parts List" provides comprehensive information on the operation and maintenance of various models of Precision Premium Ovens. Key instructions include reading the manual thoroughly, ensuring qualified personnel handle maintenance, and following proper installation procedures. The ovens feature a combined power switch and circuit breaker. The manual also includes a parts list and troubleshooting guidance.
Description:
Operating manual and parts list
Environment:
Precision Premium Ovens, 3050 Series, models:
| Models | Size | Voltage | Forced Air/Gravity |
| PR305040G (6947) - Small | 2.5 cu ft | 240V | Gravity |
| PR305040GCN (6948) - Small | 2.5 cu ft | 240V | Gravity |
| PR305040M (6949) - Small | 2.5 cu ft | 240V | Forced Air |
| PR305040MCN (6950) - Small | 2.5 cu ft | 240V | Forced Air |
| PR305045G (6951) - Small | 2.5 cu ft | 120V | Gravity |
| PR305045M (6952 )- Small | 2.5 cu ft | 120V | Forced Air |
| PR305050G (6953) - Medium | 3.8 cu ft | 240V | Gravity |
| PR305050GCN (6954) - Medium | 3.8 cu ft | 240V | Gravity |
| PR305050M (6955) - Medium | 3.8 cu ft | 240V | Forced Air |
| PR305050MCN (6956) - Medium | 3.8 cu ft | 240V | Forced Air |
| PR305055G (6957) - Medium | 3.8 cu ft | 120V | Gravity |
| PR305055M (6958) - Medium | 3.8 cu ft | 120V | Forced Air |
| PR305060G (6959) - Large | 5 cu ft | 240V | Gravity |
| PR305060GCN (6960) - Large | 5 cu ft | 240V | Gravity |
| PR305060M (6961) - Large | 5 cu ft | 240V | Forced Air |
| PR305060MCN (6962) - Large | 5 cu ft | 240V | Forced Air |
| PR305065G (6963) - Large | 5 cu ft | 120V | Gravity |
| PR305065M (6964) - Large | 5 cu ft | 120V | Forced Air |
Attachment(s):
| File | Last Modified |
|---|---|
| LT2148X2 - Rev 1 - Precision Premium Ovens, 3050 Series - Operating Manual and Parts List.pdf | December 21, 2021 |