LT2148X3 - Rev 0 - Lindberg Blue M Performance Ovens, 3055 and 3056 Series - Operating Manual and Parts List
This document provides an operating manual and parts list for Lindberg Blue M Performance Ovens. It covers features like PID Microprocessor controls, temperature ranges of 50°C to 275°C, and air circulation methods for gravity and forced air models. It includes safety instructions, maintenance guidelines, and troubleshooting steps, emphasizing the need for qualified personnel to handle internal adjustments.
Description:
Operating manual and parts list
Environment:
Lindberg Blue M Performance Ovens, 3055 and 3056 Series, models:
| Models | Size | Voltage | Forced Air/Gravity |
| LB305545G (6941) | 2.5 cu ft (Small) | 120V | Gravity |
| LB305645M (6944) | 2.5 cu ft (Small) | 120V | Forced Air |
| LB305555G (6942) | 3.8 cu ft (Medium) | 120V | Gravity |
| LB305655M (6945) | 3.8 cu ft (Medium) | 120V | Forced Air |
| LB305565G (6943) | 5 cu ft (Large) | 120V | Gravity |
| LB305665M (6946) | 5 cu ft (Large) | 120V | Forced Air |
Attachment(s):
| File | Last Modified |
|---|---|
| LT2148X3 - Rev 0 - Lindberg Blue M Performance Ovens, 3055 and 3056 Series - Operating Manual and Parts List.pdf | December 20, 2021 |