3940 Series - Troubleshooting Humidity Sensor and Control - Guide
Guide covers 3940 humidity control: Vaisala sensor sends 0–5V to Watlow controller; steam heat. Errors reflect out-of-range signals. Resolution: check 12 V supply, wiring, sensor output to restore RH.
Description:
Troubleshooting Humidity Sensor and Control for 3940
Environment:
3940 Series
Attachment(s):
| File | Last Modified |
|---|---|
| 3940 Series - Troubleshooting Humidity Sensor and Control - Guide | January 19, 2022 |