50158234 - Rev B - Thermo Scientific Environmental Chamber, Model 3907 and 3948 - Operating Manual
Description:
Operating manual
Environment:
- Thermo Scientific Environmental Chamber, Model 3907
- Thermo Scientific Environmental Chamber, Model 3948
Parameter | Unit | 3907 | 3948 |
Chamber volume | L / cu ft | 311 / 11 | 821 / 29 |
Chamber temperature range | °C (°F) | 0 - 60 (32 - 140) | 0 - 60 (32 - 140) |
Chamber humidity range | % RH | above ambient to 95 | above ambient to 95 |
Attachment(s):
File | Last Modified |
---|---|
50158234 - Rev B - Thermo Scientific Environmental Chamber, Model 3907 and 3948 - Operating Manual.pdf | December 15, 2021 |