Thermo Fisher Scientific
057-139-00 - Lab-Line Compact Low Cost Incubators - Operation Manual
The purpose of the "057-139-00 - Lab-Line Compact Low Cost Incubators - Operation Manual" is to serve as a comprehensive guide for the operation, repair, and maintenance of Lab-Line Compact Low Cost Incubators. This manual includes detailed instructions on how to use the incubators, troubleshoot common issues, and order replacement parts. It also specifies the model numbers and power requirements for the incubators.
Description: 
Operation and repair manual and parts list
Environment:
Lab-Line Compact Low Cost Incubators, model numbers and power requirements:
Model: Power Requirements:
  • 100
  • 100IDE
  • 105
  • 120
  • 125
  • 120VAC, 50/60 Hz, 100 Watts, 0.8 Amps
  • 120VAC, 50/60 Hz, 100 Watts, 0.8 Amps
  • 120VAC, 50/60 Hz, 100 Watts, 0.8 Amps
  • 120VAC, 50/60 Hz, 100 Watts, 0.8 Amps
  • 120VAC, 50/60 Hz, 100 Watts, 0.8 Amps
  • 150
  • 120VAC, 50/60 Hz, 150 Watts, 1.3 Amps
  • 150JPN
  • 100VAC, 50/60 Hz, 104 Watts, 1.0 Amps
  • 101
  • 106
  • 121
  • 126
  • 240VAC, 50/60 Hz, 100 Watts, 0.4 Amps
  • 240VAC, 50/60 Hz, 100 Watts, 0.4 Amps
  • 240VAC, 50/60 Hz, 100 Watts, 0.4 Amps
  • 240VAC, 50/60 Hz, 100 Watts, 0.4 Amps
  • 152
  • 240VAC, 50/60 Hz, 150 Watts, 0.6 Amps
  • 100JPN
  • 120JPN
  • 100VAC, 50/60 Hz, 70 Watts, 0.7 Amps
  • 100VAC, 50/60 Hz, 70 Watts, 0.7 Amps
  • 100-5JPN
  • 120-5JPN
  • 100VAC, 50/60 Hz, 100 Watts, 1.0 Amps
  • 100VAC, 50/60 Hz, 100 Watts, 1.0 Amps