057-325-00 - Lab Rotators and Low Cost Orbital Shakers - User Manual
The manual outlines safe operation, features, setup, use, and maintenance of Thermo Scientific Lab Rotators and Orbital Shakers, confirming they provide reliable mixing solutions with warranty support.
Description:
User manual
Environment:
Lab Rotators and Low Cost Orbital Shakers, model numbers:
| Models: | Electrical Characteristics: |
| 2309 | 120 VAC, 50/60 Hz, 0.4 Amps, 50 Watts |
| 2314 | 120 VAC, 50/60 Hz, 0.4 Amps, 50 Watts |
| 2345 | 120 VAC, 50/60 Hz, 0.4 Amps, 50 Watts |
| 2346 | 120 VAC, 50/60 Hz, 0.4 Amps, 50 Watts |
| 2309-1CE | 240 VAC, 50/60 Hz, 0.2 Amps, 50 Watts |
| 2314-1CE | 240 VAC, 50/60 Hz, 0.2 Amps, 50 Watts |
| 2345-1CE | 240 VAC, 50/60 Hz, 0.2 Amps, 50 Watts |
| 2346-1CE | 240 VAC, 50/60 Hz, 0.2 Amps, 50 Watts |
| 2309-1CECN | 240 VAC, 50/60 Hz, 0.2 Amps, 50 Watts |
| 2314-1CECN | 240 VAC, 50/60 Hz, 0.2 Amps, 50 Watts |
| 2345-1CECN | 240 VAC, 50/60 Hz, 0.2 Amps, 50 Watts |
| 2346-1CECN | 240 VAC, 50/60 Hz, 0.2 Amps, 50 Watts |
| 2309JPN | 100 VAC, 50/60 Hz, 0.5 Amps, 50 Watts |
| 2314JPN | 100 VAC, 50/60 Hz, 0.5 Amps, 50 Watts |
| 2345JPN | 100 VAC, 50/60 Hz, 0.5 Amps, 50 Watts |
| 2346JPN | 100 VAC, 50/60 Hz, 0.5 Amps, 50 Watts |
Attachment(s):
| File | Last Modified |
|---|---|
| 057-325-00 - Lab Rotators and Low Cost Orbital Shakers - User Manual.pdf | December 29, 2021 |