46014 - Rev C - Clinical Rotator, Model 341 - User Manual
Utilize the Model 341 Clinical Rotator for the Fisher Reagent Screen Test Kit for syphilis. Ensure the rotator operates at 100 RPM, with a timer setting up to 15 minutes. Use the rubber-matted platform to secure up to four test cards and the plastic cover to add moisture. Follow the provided instructions for unpacking, operation, and maintenance. Replacement parts and diagrams are included. No images or attachments are provided.
Description:
User manual
Environment:
Fisher Scientific, Clinical Rotator, Model 341. Catalog number 14-252.
Attachment(s):
| File | Last Modified |
|---|---|
| 46014 - Rev C - Clinical Rotator, Model 341 - User Manual.pdf | January 07, 2022 |